Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Pyridin-2-yl)phenol 97%

Supplier: AMBEED, INC
Warning

A229568-5G A229568-100MG A229568-250MG A229568-1G
103799-512EA 25 USD InStock
103799-512 103819-482 103850-382 76976-098 76976-100 77089-230
2-(Pyridin-2-yl)phenol 97%
2-(Pyridin-2-yl)phenol
Precaución: for Research Use Only


Formula: C₁₁H₉NO
MW: 171.195 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18072507
CAS Number: 33421-36-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty