Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(2-Aminoethyl)pyridine 98%

Supplier: AMBEED, INC
Danger

A359738-1G A359738-5G A359738-25G A359738-250MG
103826-234EA 11 USD InStock
103826-234 103837-242 76840-426 77278-276
3-(2-Aminoethyl)pyridine 98%
3-(2-Aminoethyl)pyridine
Precaución: for Research Use Only


Formula: C₇H₁₀N₂
Boiling Pt: 228.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00191604
CAS Number: 20173-24-4
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty