Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyrrolidin-1-ylacetic acid 97%

Supplier: AMBEED, INC
Warning

A178123-5G A178123-250MG A178123-1G A178123-25G A178123-5G
103807-298EA 27 USD InStock
103807-298 103845-638 76720-764 77083-038 77083-040 77301-542
Pyrrolidin-1-ylacetic acid 97%
Pyrrolidin-1-ylacetic acid
Precaución: for Research Use Only


Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
Boiling Pt: 233.8 °C (760 mm Hg)
Flash Pt: 95.2 °C
Storage Temperature: Ambient
MDL Number: MFCD04114075
CAS Number: 37386-15-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty