Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Boc-1,2,3,4-tetrahydroisoquinoline-6-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A141712-1G A141712-100MG A141712-250MG A141712-1G A141712-5G
103803-046EA 194 USD InStock
103803-046 77010-368 77010-370 77316-218 77316-220
N-Boc-1,2,3,4-tetrahydroisoquinoline-6-carboxylic acid 97%
N-Boc-1,2,3,4-tetrahydroisoquinoline-6-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₅H₁₉NO₄
Boiling Pt: 429.3 °C (760 mm Hg)
Flash Pt: 213.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05861548
CAS Number: 170097-67-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty