Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-tert-Butyl-1H-imidazole 95%

Supplier: AMBEED, INC
Warning

A138151-1G A138151-250MG A138151-1G A138151-5G A138151-100MG
103802-570EA 14 USD InStock
103802-570 76996-770 77315-592 77315-594 77881-086
2-tert-Butyl-1H-imidazole 95%
2-tert-Butyl-1H-imidazole
Precaución: for Research Use Only


Formula: C₇H₁₂N₂
MW: 124.19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16618480
CAS Number: 36947-69-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty