Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Thiazol-2-yl)benzaldehyde 95%

Supplier: AMBEED, INC
Warning

A122905-1G A122905-250MG A122905-1G A122905-5G
103800-304EA 65 USD InStock
103800-304 76797-484 77297-900 77297-902
2-(Thiazol-2-yl)benzaldehyde 95%
2-(Thiazol-2-yl)benzaldehyde
Precaución: for Research Use Only


Formula: C₁₀H₇NOS
Boiling Pt: 356.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06657778
CAS Number: 223575-69-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty