Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trifluoromethoxy)aniline 97%

Supplier: AMBEED, INC
Danger

A367971-5G A367971-25G A367971-10G A367971-5G
103837-444EA 5 USD InStock
103837-444 76840-686 77142-412 77306-794
2-(Trifluoromethoxy)aniline 97%
2-(Trifluoromethoxy)aniline
Precaución: for Research Use Only


Formula: C₇H₆F₃NO
Boiling Pt: 170.2 °C (760 mm Hg)
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00035959
CAS Number: 1535-75-7
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty