Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trifluoromethyl)phenothiazine 98%

Supplier: AMBEED, INC
Warning

A203511-5G A203511-25G A203511-100G A203511-10G A203511-5G
103848-128EA 5 USD InStock
103848-128 77086-260 77097-478 77097-480 77302-858
2-(Trifluoromethyl)phenothiazine 98%
2-(Trifluoromethyl)phenothiazine
Precaución: for Research Use Only


Formula: C₁₃H₈F₃NS
MW: 267.27 g/mol
Boiling Pt: 361.1 °C (760 mm Hg)
Melting Pt: 188…190 °C
Flash Pt: 210 °C
Storage Temperature: Ambient
MDL Number: MFCD00005018
CAS Number: 92-30-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty