Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trifluoromethyl)nicotinaldehyde 95%

Supplier: AMBEED, INC
Warning

A116072-1G A116072-250MG A116072-1G
103798-668EA 124 USD InStock
103798-668 103824-532 77245-934 77297-416
2-(Trifluoromethyl)nicotinaldehyde 95%
2-(Trifluoromethyl)nicotinaldehyde
Precaución: for Research Use Only


Formula: C₇H₄F₃NO
Boiling Pt: 207.9 °C (760 mm Hg)
Flash Pt: 79.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08234912
CAS Number: 116308-35-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty