Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trifluoromethyl)thiazol-5-amine 95%

Supplier: AMBEED, INC
Warning

A399513-1G A399513-100MG A399513-250MG
103827-034EA 491 USD InStock
103827-034 77240-610 77240-612
2-(Trifluoromethyl)thiazol-5-amine 95%
2-(Trifluoromethyl)thiazol-5-amine
Precaución: Keep in a dark place, sealed in dry, at 2…8°C


Formula: C₄H₃F₃N₂S
Storage Temperature: Refrigerator
MDL Number: MFCD11847091
CAS Number: 1367944-72-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty