Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trifluoroacetohydrazide 95%

Supplier: AMBEED, INC
Danger

A154083-1G A154083-5G A154083-100MG A154083-10G A154083-250MG A154083-1G A154083-25G A154083-5G
103804-602EA 8 USD InStock
103804-602 103842-782 76783-520 76783-522 76783-524 77318-474 77318-476 77318-478
Trifluoroacetohydrazide 95%
Trifluoroacetohydrazide
Precaución: for Research Use Only


Formula: C₂H₃F₃N₂O
Boiling Pt: 192.3 °C (760 mm Hg)
Melting Pt: 111 °C
Flash Pt: 70.1 °C
Storage Temperature: Freezer
MDL Number: MFCD00221440
CAS Number: 1538-08-5
UN: 1325
ADR: 4.1,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty