Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2,3,3-Tetramethylcyclopropanecarboxylic acid 95%

Supplier: AMBEED, INC
Warning

A243676-5G A243676-25G A243676-1000G A243676-100G A243676-10G A243676-5G
103851-360EA 6 USD InStock
103851-360 77090-568 77193-430 77193-432 77193-434 77304-274
2,2,3,3-Tetramethylcyclopropanecarboxylic acid 95%
2,2,3,3-Tetramethylcyclopropanecarboxylic acid
Precaución: for Research Use Only


Formula: C₈H₁₄O₂
MW: 142.20 g/mol
Boiling Pt: 204.8 °C (760 mm Hg)
Melting Pt: 120…121 °C
Flash Pt: 97.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00011538
CAS Number: 15641-58-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty