Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2'-Bipyridine-5,5'-dicarboxylic acid 97%

Supplier: AMBEED, INC
Warning

A134086-1G A134086-10G A134086-250MG A134086-1G A134086-25G A134086-5G A134086-100G
103801-970EA 9 USD InStock
103801-970 76996-548 76996-550 77314-682 77314-684 77314-686 77599-930
2,2'-Bipyridine-5,5'-dicarboxylic acid 97%
2,2'-Bipyridine-5,5'-dicarboxylic acid
Precaución: for Research Use Only


Formula: C₁₂H₈N₂O₄
MW: 244.21 g/mol
Boiling Pt: 521 °C (760 mm Hg)
Melting Pt: >360 °C
Flash Pt: 268.9 °C
Storage Temperature: Ambient
MDL Number: MFCD01318320
CAS Number: 1802-30-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty