Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Dimethyl-N-pyridin-2-ylpropionamide 98%

Supplier: AMBEED, INC
Warning

A192415-25G A192415-100G A192415-10G
103816-440EA 6 USD InStock
103816-440 103847-046 77084-914 77084-916 77084-918 77151-928 77151-930
2,2-Dimethyl-N-pyridin-2-ylpropionamide 98%
2,2-Dimethyl-N-pyridin-2-ylpropionamide
Precaución: for Research Use Only


Formula: C₁₀H₁₄N₂O
MW: 178.235 g/mol
Melting Pt: 71…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00460277
CAS Number: 86847-59-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty