Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Diphenylcyclopropanecarbonitrile 97%

Supplier: AMBEED, INC
Warning

A240881-1G A240881-5G A240881-250MG A240881-25G A240881-5G A240881-1G
103811-372EA 10 USD InStock
103811-372 103851-178 76874-410 76874-412 77090-336 77304-204
2,2-Diphenylcyclopropanecarbonitrile 97%
2,2-Diphenylcyclopropanecarbonitrile
Precaución: for Research Use Only


Formula: C₁₆H₁₃N
Boiling Pt: 374.9 °C (760 mm Hg)
Flash Pt: 141.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00182382
CAS Number: 30932-41-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty