Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Selenocystamine dihydrochloride 97%

Supplier: AMBEED, INC
Danger

A152237-1G A152237-5G A152237-10G A152237-250MG
103804-368EA 27 USD InStock
103804-368 103842-532 76719-788 76719-790
Selenocystamine dihydrochloride 97%
Selenocystamine dihydrochloride
Precaución: for Research Use Only


Formula: C₄H₁₄Cl₂N₂Se₂
Flash Pt: 152.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00035445
CAS Number: 3542-13-0
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty