Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2'-Oxybis(ethylamine) 98%

Supplier: AMBEED, INC
Danger

A308790-1G A308790-5G A308790-1G A308790-25G A308790-5G
103814-580EA 18 USD InStock
103814-580 103835-758 76838-520 76838-522 76838-524
2,2'-Oxybis(ethylamine) 98%
2,2'-Oxybis(ethylamine)
Precaución: for Research Use Only


Formula: C₄H₁₂N₂O
Boiling Pt: 185.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00466591
CAS Number: 2752-17-2
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty