Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3,3-Trimethylindolenine 98%

Supplier: AMBEED, INC
Warning

A117804-5G A117804-1000G A117804-100G A117804-10G A117804-25G A117804-500G A117804-5G
103824-826EA 6 USD InStock
103824-826 76797-180 76797-182 76797-184 76797-186 76797-188 77297-556
2,3,3-Trimethylindolenine 98%
2,3,3-Trimethylindolenine
Precaución: for Research Use Only


Formula: C₁₁H₁₃N
MW: 159.23 g/mol
Boiling Pt: 227.2 °C (760 mm Hg)
Melting Pt: 2…8 °C
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00005724
CAS Number: 1640-39-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty