Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3,4,5-Tetrahydro-1H-1,5-methanobenzo[d]azepine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A117279-1G A117279-5G A117279-100MG A117279-10G A117279-250MG A117279-1G A117279-5G
103798-860EA 45 USD InStock
103798-860 103824-734 76822-176 76822-178 76822-180 77311-520 77311-522
2,3,4,5-Tetrahydro-1H-1,5-methanobenzo[d]azepine hydrochloride 97%
2,3,4,5-Tetrahydro-1H-1,5-methanobenzo[d]azepine hydrochloride
Precaución: for Research Use Only


Formula: C₁₁H₁₄ClN
Boiling Pt: 277.9 °C (760 mm Hg)
Flash Pt: 128.9 °C
Storage Temperature: Ambient
MDL Number: MFCD11042279
CAS Number: 230615-52-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty