Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,5,7-Triazabicyclo[4.4.0]dec-5-ene 97%

Supplier: AMBEED, INC
Danger

A139355-1G A139355-10G A139355-1G A139355-25G A139355-5G
103802-730EA 9 USD InStock
103802-730 77190-330 77299-074 77315-816 77315-818
1,5,7-Triazabicyclo[4.4.0]dec-5-ene 97%
1,5,7-Triazabicyclo[4.4.0]dec-5-ene
Precaución: for Research Use Only


Formula: C₇H₁₃N₃
MW: 139.20 g/mol
Melting Pt: 125…130 °C
Flash Pt: 88.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00043003
CAS Number: 5807-14-7
UN: 1759
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty