Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tryptoline 97%

Supplier: AMBEED, INC
Warning

A101791-1G A101791-5G A101791-10G A101791-250MG A101791-25G A101791-1G A101791-5G A101791-100G
103796-138EA 12 USD InStock
103796-138 103821-826 76796-286 76796-288 76796-290 77308-722 77308-724 77878-797
Tryptoline 97%
Tryptoline
Precaución: for Research Use Only


Formula: C₁₁H₁₂N₂
Boiling Pt: 351.6 °C (760 mm Hg)
Melting Pt: 202…206 °C
Flash Pt: 166.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00004954
CAS Number: 16502-01-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty