Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Nitro-2,3,4-trifluoroaniline 98%

Supplier: AMBEED, INC
Warning

A170641-1G A170641-5G A170641-1G A170641-25G A170641-5G A170641-10G
103806-520EA 11 USD InStock
103806-520 103844-794 77081-904 77081-906 77081-908 77151-264
6-Nitro-2,3,4-trifluoroaniline 98%
6-Nitro-2,3,4-trifluoroaniline
Precaución: for Research Use Only


Formula: C₆H₃F₃N₂O₂
MW: 192.1
Melting Pt: 59…61 °C
Flash Pt: 75.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00174089
CAS Number: 148416-38-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty