Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-2,3,5,6-tetrafluoropyridine 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A385083-1G A385083-5G A385083-1G A385083-10G
103826-778EA 9 USD InStock
103826-778 103837-816 76841-158 77306-970 77684-109
4-Amino-2,3,5,6-tetrafluoropyridine 98%, Ambeed.Inc
4-Amino-2,3,5,6-tetrafluoropyridine
Precaución: For research use only.


Formula: C₅H₂F₄N₂
Boiling Pt: 265.4 °C (760 mm Hg)
Melting Pt: 85…87 °C at N/A mmHg
Flash Pt: 114.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00010855
CAS Number: 1682-20-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty