Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3,5,6-Tetrafluoroterephthalaldehyde 98%

Supplier: AMBEED, INC
Warning

A348790-1G A348790-5G A348790-250MG A348790-1G
103825-996EA 21 USD InStock
103825-996 103836-986 76840-064 77263-830 77306-598
2,3,5,6-Tetrafluoroterephthalaldehyde 98%
2,3,5,6-Tetrafluoroterephthalaldehyde
Precaución: for Research Use Only


Formula: C₈H₂F₄O₂
Boiling Pt: 252.566 °C (760 mm Hg)
Melting Pt: 130…132 °C at N/A mmHg
Flash Pt: 96.169 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08704256
CAS Number: 3217-47-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty