Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Diaminobenzamide 95%

Supplier: AMBEED, INC
Danger

A721466-5G A721466-10G A721466-250MG A721466-1G A721466-25G A721466-5G
103840-752EA 64 USD InStock
103840-752 76697-494 76697-496 76844-972 76844-974 77308-206
2,3-Diaminobenzamide 95%
2,3-Diaminobenzamide
Precaución: for Research Use Only


Formula: C₇H₉N₃O
Boiling Pt: 315 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09029906
CAS Number: 711007-44-2
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty