Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dibromobenzo[b]thiophene 98%

Supplier: AMBEED, INC
Warning

A332593-1G A332593-10G A332593-250MG A332593-1G A332593-25G A332593-5G
103825-612EA 9 USD InStock
103825-612 103836-578 76828-360 76828-362 76839-576 76839-578 76839-580
2,3-Dibromobenzo[b]thiophene 98%
2,3-Dibromobenzo[b]thiophene
Precaución: for Research Use Only


Formula: C₈H₄Br₂S
Melting Pt: 58…62 °C
Flash Pt: 162.5 °C
Storage Temperature: Ambient
MDL Number: MFCD06798119
CAS Number: 6287-82-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty