Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dichloro-6-nitroaniline 97%

Supplier: AMBEED, INC
Warning

A142572-1G A142572-250MG A142572-25G A142572-1G A142572-5G
103803-186EA 40 USD InStock
103803-186 77095-478 77095-480 77316-410 77316-412
2,3-Dichloro-6-nitroaniline 97%
2,3-Dichloro-6-nitroaniline
Precaución: for Research Use Only


Formula: C₆H₄Cl₂N₂O₂
Boiling Pt: 323.5 °C (760 mm Hg)
Melting Pt: 166.7 °C
Flash Pt: 149.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00110529
CAS Number: 65078-77-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty