Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dichloropyrazine 97%

Supplier: AMBEED, INC
Warning

A424825-1000G A424825-25G A424825-100G A424825-10G
103810-322EA 233 USD InStock
103810-322 76842-146 77291-460 77291-462
2,3-Dichloropyrazine 97%
2,3-Dichloropyrazine
Precaución: for Research Use Only


Formula: C₄H₂Cl₂N₂
Boiling Pt: 190.6 °C (760 mm Hg)
Melting Pt: 22.5…24.5 °C
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00040964
CAS Number: 4858-85-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty