Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dichloropyridin-4-amine 98%

Supplier: AMBEED, INC
Warning

A265712-1G A265712-5G A265712-100MG A265712-250MG A265712-5G A265712-1G
103812-748EA 68 USD InStock
103812-748 103852-644 77001-030 77001-032 77092-346 77304-930
2,3-Dichloropyridin-4-amine 98%
2,3-Dichloropyridin-4-amine
Precaución: for Research Use Only


Formula: C₅H₄Cl₂N₂
Storage Temperature: Ambient
MDL Number: MFCD01646113
CAS Number: 184416-83-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty