Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Difluoro-4-hydroxybenzonitrile 98%

Supplier: AMBEED, INC
Warning

A392329-1G A392329-5G A392329-1G A392329-5G
103826-900EA 39 USD InStock
103826-900 103837-934 77307-006 77307-008
2,3-Difluoro-4-hydroxybenzonitrile 98%
2,3-Difluoro-4-hydroxybenzonitrile
Precaución: for Research Use Only


Formula: C₇H₃F₂NO
Boiling Pt: 260.8 °C (760 mm Hg)
Melting Pt: 145…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00269596
CAS Number: 126162-38-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty