Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Difluoroaniline 98%

Supplier: AMBEED, INC
Warning

A159707-5G A159707-100G A159707-500G A159707-25G A159707-5G A159707-10G
103843-500EA 14 USD InStock
103843-500 77286-416 77286-418 77319-398 77319-400 77615-300
2,3-Difluoroaniline 98%
2,3-Difluoroaniline
Precaución: for Research Use Only


Formula: C₆H₅F₂N
Boiling Pt: 176.6 °C (760 mm Hg)
Flash Pt: 71.11 °C
Storage Temperature: Ambient
MDL Number: MFCD00010298
CAS Number: 4519-40-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty