Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Difluorobenzoic acid 97%

Supplier: AMBEED, INC
Warning

A138736-1G A138736-100G A138736-10G A138736-500G A138736-1G A138736-25G
103802-656EA 5 USD InStock
103802-656 77137-234 77137-236 77137-238 77299-010 77315-708
2,3-Difluorobenzoic acid 97%
2,3-Difluorobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₄F₂O₂
MW: 158.1 g/mol
Boiling Pt: 248.1 °C (760 mm Hg)
Melting Pt: 163…165 °C
Flash Pt: 103.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00010267
CAS Number: 4519-39-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty