Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dihydro-1H-inden-1-amine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A238807-5G A238807-25G A238807-100G A238807-10G A238807-5G
103851-042EA 8 USD InStock
103851-042 77090-176 77113-196 77113-198 77304-122
2,3-Dihydro-1H-inden-1-amine hydrochloride 95%
2,3-Dihydro-1H-inden-1-amine hydrochloride
Precaución: for Research Use Only


Formula: C₉H₁₂ClN
Melting Pt: 206…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00054294
CAS Number: 70146-15-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty