Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dihydro-4-benzofuranamine 97%

Supplier: AMBEED, INC
Warning

A183120-1G A183120-5G A183120-1G A183120-5G A183120-100MG A183120-10G A183120-250MG
103815-608EA 83 USD InStock
103815-608 103846-170 77083-758 77083-760 77248-326 77248-328 77248-330
2,3-Dihydro-4-benzofuranamine 97%
2,3-Dihydro-4-benzofuranamine
Precaución: for Research Use Only


Formula: C₈H₉NO
Boiling Pt: 268.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06659470
CAS Number: 61090-37-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty