Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dihydroxybenzonitrile 98%

Supplier: AMBEED, INC
Warning

A131212-1G A131212-10G A131212-250MG A131212-1G A131212-25G A131212-5G
103801-542EA 21 USD InStock
103801-542 77246-544 77246-546 77314-014 77314-016 77314-018
2,3-Dihydroxybenzonitrile 98%
2,3-Dihydroxybenzonitrile
Precaución: for Research Use Only


Formula: C₇H₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD02261930
CAS Number: 67984-81-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty