Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dihydroxy-N,N,N-trimethylpropan-1-aminium chloride 95%

Supplier: AMBEED, INC
Warning

A158624-1G A158624-5G A158624-250MG A158624-1G A158624-5G
103805-142EA 19 USD InStock
103805-142 103843-358 77235-746 77319-220 77319-222
2,3-Dihydroxy-N,N,N-trimethylpropan-1-aminium chloride 95%
2,3-Dihydroxy-N,N,N-trimethylpropan-1-aminium chloride
Precaución: for Research Use Only


Formula: C₆H₁₆ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD01668726
CAS Number: 34004-36-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty