Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxy-2-pyridone 97%

Supplier: AMBEED, INC
Warning

A145178-1G A145178-5G A145178-10G A145178-1G A145178-25G A145178-5G
103803-498EA 5 USD InStock
103803-498 103841-580 77285-904 77316-854 77316-856 77316-858
3-Hydroxy-2-pyridone 97%
3-Hydroxy-2-pyridone
Precaución: for Research Use Only


Formula: C₅H₅NO₂
MW: 111.1 g/mol
Boiling Pt: 315
Melting Pt: 245 °C
Storage Temperature: Ambient
MDL Number: MFCD00006271
CAS Number: 16867-04-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty