Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dimethoxyphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A247558-1G A247558-5G A247558-1G A247558-25G A247558-5G A247558-100G A247558-10G
103811-780EA 5 USD InStock
103811-780 103851-616 77090-954 77090-956 77090-958 77193-552 77193-554
2,3-Dimethoxyphenylboronic acid 98%
2,3-Dimethoxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₈H₁₁BO₄
Boiling Pt: 347.9 °C (760 mm Hg)
Melting Pt: 67…71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02683112
CAS Number: 40972-86-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty