Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dimethylpyrazine 98%

Supplier: AMBEED, INC
Danger

A618688-5G A618688-100G A618688-10G A618688-25G
103840-198EA 5 USD InStock
103840-198 76775-354 76775-356 77307-952
2,3-Dimethylpyrazine 98%
2,3-Dimethylpyrazine
Precaución: for Research Use Only


Formula: C₆H₈N₂
MW: 108.14
Boiling Pt: 158.1 °C (760 mm Hg)
Melting Pt: 11…13 °C
Density: 1
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00006144
CAS Number: 5910-89-4
UN: 1993
ADR: 3 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty