Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,4,5,5-Pentamethyl-1,3,2-dioxaborolane 98%

Supplier: AMBEED, INC
Danger

A789000-5G A789000-100G A789000-10G A789000-1G A789000-5G A789000-25G
103841-008EA 11 USD InStock
103841-008 76832-216 76832-218 76845-260 76845-262 77308-336
2,4,4,5,5-Pentamethyl-1,3,2-dioxaborolane 98%
2,4,4,5,5-Pentamethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only


Formula: C₇H₁₅BO₂
Storage Temperature: Freezer
MDL Number: MFCD09027082
CAS Number: 94242-85-0
UN: 3272
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty