Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,6-Trichloro-5-cyanopyrimidine 98%

Supplier: AMBEED, INC
Warning

A425559-1G A425559-5G A425559-10G A425559-250MG A425559-1G A425559-25G A425559-5G
103827-462EA 33 USD InStock
103827-462 103838-512 77252-384 77252-386 77307-230 77307-232 77307-234
2,4,6-Trichloro-5-cyanopyrimidine 98%
2,4,6-Trichloro-5-cyanopyrimidine
Precaución: Inert atmosphere, 2…8 °C.


Formula: C₅Cl₃N₃
Boiling Pt: 320.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07636745
CAS Number: 3029-64-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty