Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,6-Trifluorobenzonitrile 98%

Supplier: AMBEED, INC
Warning

A107724-1G A107724-5G A107724-10G A107724-25G A107724-500G A107724-1G A107724-5G
103797-238EA 5 USD InStock
103797-238 103823-006 77107-780 77107-782 77107-784 77283-636 77309-862
2,4,6-Trifluorobenzonitrile 98%
2,4,6-Trifluorobenzonitrile
Precaución: for Research Use Only


Formula: C₇H₂F₃N
Boiling Pt: 228.5 °C (760 mm Hg)
Melting Pt: 57…61 °C
Flash Pt: 64.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00042399
CAS Number: 96606-37-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty