Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,6-Trihydroxybenzaldehyde 95%

Supplier: AMBEED, INC
Warning

A203770-1G A203770-5G A203770-25G A203770-5G A203770-100G A203770-10G A203770-250MG A203770-1G
103817-490EA 7 USD InStock
103817-490 103848-150 77086-284 77086-286 77139-406 77139-408 77139-410 77302-872
2,4,6-Trihydroxybenzaldehyde 95%
2,4,6-Trihydroxybenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₆O₄
Boiling Pt: 319.1 °C (760 mm Hg)
Melting Pt: 195 °C
Flash Pt: 161 °C
Storage Temperature: Ambient
MDL Number: MFCD00003329
CAS Number: 487-70-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty