Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,6-Triisopropylbenzenesulfonyl hydrazide 97%

Supplier: AMBEED, INC
Danger

A144207-1G A144207-5G A144207-10G A144207-1G A144207-5G
103803-376EA 6 USD InStock
103803-376 103841-452 77272-614 77316-690 77316-692
2,4,6-Triisopropylbenzenesulfonyl hydrazide 97%
2,4,6-Triisopropylbenzenesulfonyl hydrazide
Precaución: for Research Use Only


Formula: C₁₅H₂₆N₂O₂S
Boiling Pt: 396.6 °C (760 mm Hg)
Melting Pt: 110…112 °C
Flash Pt: 193.7 °C
Storage Temperature: Freezer
MDL Number: MFCD00014750
CAS Number: 39085-59-1
UN: 1325
ADR: 4.1,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty