Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,6-Trimethylbenzoic acid 98%

Supplier: AMBEED, INC
Warning

A271136-5G A271136-10G A271136-5G A271136-500G
103809-888EA 357 USD InStock
103809-888 103852-944 76977-188 77092-716 77881-737
2,4,6-Trimethylbenzoic acid 98%
2,4,6-Trimethylbenzoic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₂
Boiling Pt: 296.6 °C (760 mm Hg)
Melting Pt: 152…155 °C
Flash Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00002481
CAS Number: 480-63-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty