Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dibromo-1,3,5-trifluorobenzene 97%

Supplier: AMBEED, INC
Warning

A171120-1G A171120-5G A171120-100MG A171120-250MG A171120-1G A171120-25G
103806-594EA 26 USD InStock
103806-594 103844-868 77138-416 77138-418 77301-096 77301-098
2,4-Dibromo-1,3,5-trifluorobenzene 97%
2,4-Dibromo-1,3,5-trifluorobenzene
Precaución: for Research Use Only


Formula: C₆HBr₂F₃
Boiling Pt: 85…90 °C (20 mm Hg)
Flash Pt: 66 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11845962
CAS Number: 363-69-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty