Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloro-1,8-naphthyridine 97%

Supplier: AMBEED, INC
Warning

A190384-1G A190384-5G A190384-1G A190384-25G A190384-5G A190384-10G A190384-250MG
103816-290EA 24 USD InStock
103816-290 103846-892 77084-684 77084-686 77084-688 77248-554 77248-556
2,4-Dichloro-1,8-naphthyridine 97%
2,4-Dichloro-1,8-naphthyridine
Precaución: for Research Use Only


Formula: C₈H₄Cl₂N₂
Boiling Pt: 299.1 °C (760 mm Hg)
Flash Pt: 163 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00234380
CAS Number: 59514-89-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty