Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine 98%

Supplier: AMBEED, INC
Warning

A158796-1G A158796-5G A158796-10G A158796-1G A158796-25G
103805-174EA 7 USD InStock
103805-174 103843-388 76973-434 77319-240 77319-242
2,4-Dichloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine 98%
2,4-Dichloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine
Precaución: for Research Use Only


Formula: C₆H₂Cl₂IN₃
MW: 313.911 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13189364
CAS Number: 1012785-51-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty