Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloro-5-sulfamoylbenzoic acid 98%

Supplier: AMBEED, INC
Warning

A123232-100G A123232-10G A123232-25G A123232-500G
103808-270EA 154 USD InStock
103808-270 76766-708 76766-710 76766-712 76766-714
2,4-Dichloro-5-sulfamoylbenzoic acid 98%
2,4-Dichloro-5-sulphamoylbenzoic acid
Precaución: for Research Use Only


Formula: C₇H₅Cl₂NO₄S
MW: 270.09 g/mol
Boiling Pt: 503.8 °C (760 mm Hg)
Melting Pt: 230…234 °C
Flash Pt: 258.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00007931
CAS Number: 2736-23-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty