Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloro-6-phenyl-1,3,5-triazine 98%

Supplier: AMBEED, INC
Warning

A456413-1G A456413-5G A456413-25G A456413-5G A456413-10G
103827-860EA 9 USD InStock
103827-860 103838-936 76842-790 76842-792 77004-374
2,4-Dichloro-6-phenyl-1,3,5-triazine 98%
2,4-Dichloro-6-phenyl-1,3,5-triazine
Precaución: for Research Use Only


Formula: C₉H₅Cl₂N₃
MW: 226.0600 g/mol
Boiling Pt: 427.5 °C (760 mm Hg)
Flash Pt: 244.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00047340
CAS Number: 1700-02-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty